2,4-Difluoro-3-methoxybenzenesulphonyl chloride
Lieferant: Apollo Scientific
PC303227-1G
APOSPC303227-1GEA
899
EUR
APOSPC303227-1G
2,4-Difluoro-3-methoxybenzenesulphonyl chloride
2,4-Difluoro-3-methoxybenzenesulphonyl chloride
Formel:
C₇H₅ClF₂O₃S Molecular Weight: 242,628 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28054212 CAS-Nummer: 1706458-65-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...