2-Methyl-5-(trifluoromethoxy)benzenesulphonyl chloride 97%

Lieferant: Apollo Scientific

PC303230-1G PC303230-250MG
APOSPC303230-1GEA 0 EUR
APOSPC303230-1G APOSPC303230-250MG
2-Methyl-5-(trifluoromethoxy)benzenesulphonyl chloride 97%
2-Methyl-5-(trifluoromethoxy)benzenesulphonyl chloride
Formel: C₈H₆ClF₃O₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD18399733
CAS-Nummer: 1261852-46-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR