3-Chloro-4-ethoxy-5-fluorobenzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303233-1G
APOSPC303233-1GEA 899 EUR
APOSPC303233-1G
3-Chloro-4-ethoxy-5-fluorobenzenesulphonyl chloride
3-Chloro-4-ethoxy-5-fluorobenzenesulphonyl chloride
Formel: C₈H₇Cl₂FO₃S
Molecular Weight: 273,109 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28054225
CAS-Nummer: 1706430-92-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR