4-Chloro-2,6-difluorobenzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303234-1G
APOSPC303234-1GEA 899 EUR
APOSPC303234-1G
4-Chloro-2,6-difluorobenzenesulphonyl chloride
4-Chloro-2,6-difluorobenzenesulphonyl chloride
Formel: C₆H₂Cl₂F₂O₂S
Molecular Weight: 247,047 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12922627
CAS-Nummer: 1208077-94-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR