2-Fluoro-5-methoxy-3-(trifluoromethyl)benzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303238-1G
APOSPC303238-1GEA 956 EUR
APOSPC303238-1G
2-Fluoro-5-methoxy-3-(trifluoromethyl)benzenesulphonyl chloride
2-Fluoro-5-methoxy-3-(trifluoromethyl)benzenesulphonyl chloride
Formel: C₈H₅ClF₄O₃S
Molecular Weight: 292,635 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28054243
CAS-Nummer: 1706435-15-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR