4-Methyl-2,5-bis(trifluoromethyl)benzoic acid 97%

Lieferant: Apollo Scientific

PC303239-1G PC303239-250MG
APOSPC303239-1GEA 0 EUR
APOSPC303239-1G APOSPC303239-250MG
4-Methyl-2,5-bis(trifluoromethyl)benzoic acid 97%
4-Methyl-2,5-bis(trifluoromethyl)benzoic acid
Formel: C₁₀H₆F₆O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD27986831
CAS-Nummer: 1706436-17-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR