2-Chloro-6-fluoro-3-methoxybenzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303245-1G
APOSPC303245-1GEA 0 EUR
APOSPC303245-1G
2-Chloro-6-fluoro-3-methoxybenzenesulphonyl chloride
2-Chloro-6-fluoro-3-methoxybenzenesulphonyl chloride
Formel: C₇H₅Cl₂FO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD28054219
CAS-Nummer: 1706430-87-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR