2-Methoxy-4,6-bis(trifluoromethyl)benzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303247-1G
APOSPC303247-1GEA 0 EUR
APOSPC303247-1G
2-Methoxy-4,6-bis(trifluoromethyl)benzenesulphonyl chloride
2-Methoxy-4,6-bis(trifluoromethyl)benzenesulphonyl chloride
Formel: C₉H₅ClF₆O₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD28054227
CAS-Nummer: 1706439-13-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR