3-Bromo-5-(trifluoromethoxy)benzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303248-1G
APOSPC303248-1GEA 1640 EUR
APOSPC303248-1G
3-Bromo-5-(trifluoromethoxy)benzenesulphonyl chloride
3-Bromo-5-(trifluoromethoxy)benzenesulphonyl chloride
Formel: C₇H₃BrClF₃O₃S
Molecular Weight: 339,514 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28054230
CAS-Nummer: 1706446-78-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR