3-Chloro-5-fluoro-4-methoxybenzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303249-1G
APOSPC303249-1GEA 1020 EUR
APOSPC303249-1G
3-Chloro-5-fluoro-4-methoxybenzenesulphonyl chloride
3-Chloro-5-fluoro-4-methoxybenzenesulphonyl chloride
Formel: C₇H₅Cl₂FO₃S
Molecular Weight: 259,082 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28054214
CAS-Nummer: 1706458-23-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR