3-Ethoxy-2,6-difluorobenzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303250-1G
APOSPC303250-1GEA 1020 EUR
APOSPC303250-1G
3-Ethoxy-2,6-difluorobenzenesulphonyl chloride
3-Ethoxy-2,6-difluorobenzenesulphonyl chloride
Formel: C₈H₇ClF₂O₃S
Molecular Weight: 256,654 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24284208
CAS-Nummer: 1552934-62-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR