4-Ethoxy-2,3-difluorobenzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303251-1G
APOSPC303251-1GEA 1020 EUR
APOSPC303251-1G
4-Ethoxy-2,3-difluorobenzenesulphonyl chloride
4-Ethoxy-2,3-difluorobenzenesulphonyl chloride
Formel: C₈H₇ClF₂O₃S
Molecular Weight: 256,654 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17282233
CAS-Nummer: 293299-90-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR