3-Bromo-5-(pentafluorosulphur)benzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303255-1G
APOSPC303255-1GEA 1080 EUR
APOSPC303255-1G
3-Bromo-5-(pentafluorosulphur)benzenesulphonyl chloride
3-Bromo-5-(pentafluorosulphur)benzenesulphonyl chloride
Formel: C₆H₃BrClF₅O₂S₂
Molecular Weight: 381,566 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28054233
CAS-Nummer: 1706453-16-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR