2,3-Difluoro-5-methoxy-4-methylphenylacetic acid 98%

Lieferant: Apollo Scientific

PC303256-1G PC303256-250MG
APOSPC303256-1GEA 1160 EUR
APOSPC303256-1G APOSPC303256-250MG
2,3-Difluoro-5-methoxy-4-methylphenylacetic acid 98%
2,3-Difluoro-5-methoxy-4-methylphenylacetic acid
Formel: C₁₀H₁₀F₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD27986854
CAS-Nummer: 1706430-83-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR