3-Ethoxy-2,4-difluorobenzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303258-1G
APOSPC303258-1GEA 1210 EUR
APOSPC303258-1G
3-Ethoxy-2,4-difluorobenzenesulphonyl chloride
3-Ethoxy-2,4-difluorobenzenesulphonyl chloride
Formel: C₈H₇ClF₂O₃S
Molecular Weight: 256,654 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28054223
CAS-Nummer: 1706439-10-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR