4-Ethoxy-3,5-difluorobenzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303260-1G
APOSPC303260-1GEA 1250 EUR
APOSPC303260-1G
4-Ethoxy-3,5-difluorobenzenesulphonyl chloride
4-Ethoxy-3,5-difluorobenzenesulphonyl chloride
Formel: C₈H₇ClF₂O₃S
Molecular Weight: 256,654 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24076694
CAS-Nummer: 1519155-45-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR