4-Fluoro-3-methyl-5-(trifluoromethyl)benzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303261-1G
APOSPC303261-1GEA 1250 EUR
APOSPC303261-1G
4-Fluoro-3-methyl-5-(trifluoromethyl)benzenesulphonyl chloride
4-Fluoro-3-methyl-5-(trifluoromethyl)benzenesulphonyl chloride
Formel: C₈H₅ClF₄O₂S
Molecular Weight: 276,636 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28054246
CAS-Nummer: 1706458-75-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR