2-Fluoro-5-(pentafluorosulphur)benzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303262-1G
APOSPC303262-1GEA 1280 EUR
APOSPC303262-1G
2-Fluoro-5-(pentafluorosulphur)benzenesulphonyl chloride
2-Fluoro-5-(pentafluorosulphur)benzenesulphonyl chloride
Formel: C₆H₃ClF₆O₂S₂
Molecular Weight: 320,66 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28054234
CAS-Nummer: 1706447-01-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR