2-Methyl-5-(pentafluorosulphur)benzenesulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

PC303263-1G
APOSPC303263-1GEA 1280 EUR
APOSPC303263-1G
2-Methyl-5-(pentafluorosulphur)benzenesulphonyl chloride
2-Methyl-5-(pentafluorosulphur)benzenesulphonyl chloride
Formel: C₇H₆ClF₅O₂S₂
Molecular Weight: 316,696 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28054235
CAS-Nummer: 1706458-82-3
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR