4-Methoxy-3-methyl-5-(trifluoromethyl)benzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303264-1G
APOSPC303264-1GEA 1400 EUR
APOSPC303264-1G
4-Methoxy-3-methyl-5-(trifluoromethyl)benzenesulphonyl chloride
4-Methoxy-3-methyl-5-(trifluoromethyl)benzenesulphonyl chloride
Formel: C₉H₈ClF₃O₃S
Molecular Weight: 288,671 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28054248
CAS-Nummer: 1706435-16-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR