3-Bromo-2-methyl-5-(pentafluorosulphur)benzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303265-1G
APOSPC303265-1GEA 1510 EUR
APOSPC303265-1G
3-Bromo-2-methyl-5-(pentafluorosulphur)benzenesulphonyl chloride
3-Bromo-2-methyl-5-(pentafluorosulphur)benzenesulphonyl chloride
Formel: C₇H₅BrClF₅O₂S₂
Molecular Weight: 395,592 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28054247
CAS-Nummer: 1706431-07-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR