4-Chloro-3-ethoxy-2-fluorobenzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303268-1G
APOSPC303268-1GEA 1510 EUR
APOSPC303268-1G
4-Chloro-3-ethoxy-2-fluorobenzenesulphonyl chloride
4-Chloro-3-ethoxy-2-fluorobenzenesulphonyl chloride
Formel: C₈H₇Cl₂FO₃S
Molecular Weight: 273,109 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28054241
CAS-Nummer: 1706430-95-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR