3-Chloro-5-(pentafluorosulphur)benzenesulphonyl chloride
Lieferant: Apollo Scientific
PC303270-1G
APOSPC303270-1GEA
1630
EUR
APOSPC303270-1G
3-Chloro-5-(pentafluorosulphur)benzenesulphonyl chloride
3-Chloro-5-(pentafluorosulphur)benzenesulphonyl chloride
Formel:
C₆H₃Cl₂F₅O₂S₂ Molecular Weight: 337,115 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28054236 CAS-Nummer: 1706462-61-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...