4-Butoxy-2,6-difluorobenzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303271-1G
APOSPC303271-1GEA 1630 EUR
APOSPC303271-1G
4-Butoxy-2,6-difluorobenzenesulphonyl chloride
4-Butoxy-2,6-difluorobenzenesulphonyl chloride
Formel: C₁₀H₁₁ClF₂O₃S
Molecular Weight: 284,707 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28054242
CAS-Nummer: 1706431-01-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR