3-Methyl-5-(pentafluorosulphur)benzyl alcohol 97%

Lieferant: Apollo Scientific

PC303285-1G
APOSPC303285-1GEA 0 EUR
APOSPC303285-1G
3-Methyl-5-(pentafluorosulphur)benzyl alcohol 97%
3-Methyl-5-(pentafluorosulphur)benzyl alcohol
Formel: C₈H₉F₅OS
Lagertemperatur: Raumtemperatur
MDL: MFCD29045644
CAS-Nummer: 1858256-52-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR