6-Ethoxy-2,3,4-trifluorobenzoic acid 97%

Lieferant: Apollo Scientific

PC303287-1G
APOSPC303287-1GEA 405 EUR
APOSPC303287-1G
6-Ethoxy-2,3,4-trifluorobenzoic acid 97%
6-Ethoxy-2,3,4-trifluorobenzoic acid
Formel: C₉H₇F₃O₃
Molecular Weight: 220,145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29277475
CAS-Nummer: 1980050-06-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR