5-Methyl-2-(trifluoromethoxy)benzenesulphonamide 97%

Lieferant: Apollo Scientific

PC303295-1G PC303295-250MG
APOSPC303295-1GEA 1830 EUR
APOSPC303295-1G APOSPC303295-250MG
5-Methyl-2-(trifluoromethoxy)benzenesulphonamide 97%
5-Methyl-2-(trifluoromethoxy)benzenesulphonamide
Formel: C₈H₈F₃NO₃S
Molecular Weight: 255.214 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18399730
CAS-Nummer: 1261683-36-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR