3,6-Difluoro-2-methoxyphenylacetonitrile 97%

Lieferant: Apollo Scientific

PC303316-1G
APOSPC303316-1GEA 290 EUR
APOSPC303316-1G
3,6-Difluoro-2-methoxyphenylacetonitrile 97%
3,6-Difluoro-2-methoxyphenylacetonitrile
Formel: C₉H₇F₂NO
Molecular Weight: 183.155 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18395261
CAS-Nummer: 1261760-73-9
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR