3,6-Difluoro-2-methoxybenzoyl chloride 98%

Lieferant: Apollo Scientific

PC303318-5G PC303318-1G
APOSPC303318-5GEA 0 EUR
APOSPC303318-5G APOSPC303318-1G
3,6-Difluoro-2-methoxybenzoyl chloride 98%
3,6-Difluoro-2-methoxybenzoyl chloride
Formel: C₈H₅ClF₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18379811
CAS-Nummer: 1261825-37-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR