4-Chloro-2-fluoro-5-methylphenylacetonitrile 97%

Lieferant: Apollo Scientific

PC303323-1G PC303323-5G
APOSPC303323-1GEA 310 EUR
APOSPC303323-1G APOSPC303323-5G
4-Chloro-2-fluoro-5-methylphenylacetonitrile 97%
4-Chloro-2-fluoro-5-methylphenylacetonitrile
Formel: C₉H₇ClFN
Molecular Weight: 183.61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20483139
CAS-Nummer: 1242314-99-3
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR