4-Chloro-2-fluoro-5-methylbenzoic acid 97%

Lieferant: Apollo Scientific

PC303325-1G PC303325-250MG
APOSPC303325-1GEA 299 EUR
APOSPC303325-1G APOSPC303325-250MG
4-Chloro-2-fluoro-5-methylbenzoic acid 97%
4-Chloro-2-fluoro-5-methylbenzoic acid
Formel: C₈H₆ClFO₂
Molecular Weight: 188.583 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20660427
CAS-Nummer: 177211-31-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR