4-Chloro-2-fluoro-5-methylbenzenesulphonamide
Lieferant: Apollo Scientific
PC303330-1G
APOSPC303330-1GEA
563
EUR
APOSPC303330-1G
4-Chloro-2-fluoro-5-methylbenzenesulphonamide
4-Chloro-2-fluoro-5-methylbenzenesulphonamide
Formel:
C₇H₇ClFNO₂S Molecular Weight: 223,652 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28751802 CAS-Nummer: 1805525-24-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...