2-Chloro-4-fluoro-5-methylphenylacetonitrile 97%

Lieferant: Apollo Scientific

PC303340-1G
APOSPC303340-1GEA 316 EUR
APOSPC303340-1G
2-Chloro-4-fluoro-5-methylphenylacetonitrile 97%
2-Chloro-4-fluoro-5-methylphenylacetonitrile
Formel: C₉H₇ClFN
Molecular Weight: 183,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09923912
CAS-Nummer: 1000513-29-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR