2-Chloro-4-fluoro-5-methylbenzoic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC303342-5G
PC303342-1G
APOSPC303342-5GEA
417
EUR
APOSPC303342-5G
APOSPC303342-1G
2-Chloro-4-fluoro-5-methylbenzoic acid 97%
2-Chloro-4-fluoro-5-methylbenzoic acid
Formel:
C₈H₆ClFO₂ Molecular Weight: 188.583 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20658460 CAS-Nummer: 208165-96-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...