2-Chloro-4-fluoro-5-methylbenzoyl chloride

Lieferant: Apollo Scientific

PC303343-5G
APOSPC303343-5GEA 743 EUR
APOSPC303343-5G
2-Chloro-4-fluoro-5-methylbenzoyl chloride
2-Chloro-4-fluoro-5-methylbenzoyl chloride
Formel: C₈H₅Cl₂FO
Molecular Weight: 207,029 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28751941
CAS-Nummer: 1807027-18-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR