2-Chloro-4-fluoro-5-methylbenzylamine

Lieferant: Apollo Scientific

PC303348-1G PC303348-5G
APOSPC303348-1GEA 339 EUR
APOSPC303348-1G APOSPC303348-5G
2-Chloro-4-fluoro-5-methylbenzylamine
2-Chloro-4-fluoro-5-methylbenzylamine
Formel: C₈H₉ClFN
Molecular Weight: 173.615 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21602108
CAS-Nummer: 1365968-65-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR