2-Chloro-4-fluoro-5-methylbenzylamine
Lieferant: Apollo Scientific
PC303348-1G
PC303348-5G
APOSPC303348-1GEA
339
EUR
APOSPC303348-1G
APOSPC303348-5G
2-Chloro-4-fluoro-5-methylbenzylamine
2-Chloro-4-fluoro-5-methylbenzylamine
Formel:
C₈H₉ClFN Molecular Weight: 173.615 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD21602108 CAS-Nummer: 1365968-65-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...