3,4,5-Trifluorbenzolsulfonamid 97%
Lieferant: Apollo Scientific
PC303352-1G
APOSPC303352-1GEA
40.6
EUR
APOSPC303352-1G
3,4,5-Trifluorbenzolsulfonamid 97%
3,4,5-Trifluorbenzolsulfonamid
Formel:
C₆H₄F₃NO₂S Molecular Weight: 211,16 g/mol Schmelzpunkt: 124…126 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09800668 CAS-Nummer: 913472-55-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...