5-Chloro-2,4-difluorophenylacetonitrile 97%

Lieferant: Apollo Scientific

Synonyme: 2-(5-Chloro-2,4-difluorophenyl)acetonitrile

PC303356-1G
APOSPC303356-1GEA 74.1 EUR
APOSPC303356-1G
5-Chloro-2,4-difluorophenylacetonitrile 97%
5-Chloro-2,4-difluorophenylacetonitrile
Formel: C₈H₄ClF₂N
Molecular Weight: 187,574 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28402683
CAS-Nummer: 1429422-26-3
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR