5-Chloro-2,4-difluorophenylacetic acid 95%

Lieferant: Apollo Scientific

PC303357-1G
APOSPC303357-1GEA 261 EUR
APOSPC303357-1G
5-Chloro-2,4-difluorophenylacetic acid 95%
5-Chloro-2,4-difluorophenylacetic acid
Formel: C₈H₅ClF₂O₂
Molecular Weight: 206,574 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18415640
CAS-Nummer: 1261632-58-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR