5-Chloro-2,4-difluorocinnamic acid

Lieferant: Apollo Scientific

PC303358-5G
APOSPC303358-5GEA 586 EUR
APOSPC303358-5G
5-Chloro-2,4-difluorocinnamic acid
5-Chloro-2,4-difluorocinnamic acid
Formel: C₉H₅ClF₂O₂
Molecular Weight: 218,585 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28750524
CAS-Nummer: 695187-30-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR