5-Chloro-2,4-difluorobenzoyl chloride
Lieferant: Apollo Scientific
PC303360-5G
APOSPC303360-5GEA
294
EUR
APOSPC303360-5G
5-Chloro-2,4-difluorobenzoyl chloride
5-Chloro-2,4-difluorobenzoyl chloride
Formel:
C₇H₂Cl₂F₂O Molecular Weight: 210,993 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18415575 CAS-Nummer: 1261734-09-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...