2-Bromo-4-chloro-5-fluorobenzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303363-1G
APOSPC303363-1GEA 788 EUR
APOSPC303363-1G
2-Bromo-4-chloro-5-fluorobenzenesulphonyl chloride
2-Bromo-4-chloro-5-fluorobenzenesulphonyl chloride
Formel: C₆H₂BrCl₂FO₂S
Molecular Weight: 307,952 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05664917
CAS-Nummer: 874801-47-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR