4,6-Dichloro-2-fluoro-3-methoxyphenylacetonitrile 97%

Lieferant: Apollo Scientific
Gefahr

PC303370-1G
APOSPC303370-1GEA 0 EUR
APOSPC303370-1G
4,6-Dichloro-2-fluoro-3-methoxyphenylacetonitrile 97%
4,6-Dichloro-2-fluoro-3-methoxyphenylacetonitrile
Formel: C₉H₆Cl₂FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD30830090
CAS-Nummer: 2149598-64-9
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR