4,6-Dichloro-2-fluoro-3-methoxybenzylamine

Lieferant: Apollo Scientific

PC303372-1G PC303372-5G
APOSPC303372-1GEA 0 EUR
APOSPC303372-1G APOSPC303372-5G
4,6-Dichloro-2-fluoro-3-methoxybenzylamine
4,6-Dichloro-2-fluoro-3-methoxybenzylamine
Formel: C₈H₈Cl₂FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD30726940
CAS-Nummer: 2149590-99-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR