5-Fluoro-2-(trifluoromethoxy)-DL-phenylalanine

Lieferant: Apollo Scientific

PC303373-1G PC303373-5G
APOSPC303373-1GEA 0 EUR
APOSPC303373-1G APOSPC303373-5G
5-Fluoro-2-(trifluoromethoxy)-DL-phenylalanine
5-Fluoro-2-(trifluoromethoxy)-DL-phenylalanine
Formel: C₁₀H₉F₄NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD32640048
CAS-Nummer: 1703955-84-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR