3-Chloro-2-fluoro-4-(trifluoromethyl)aniline 98%

Lieferant: Apollo Scientific

PC303375-1G
APOSPC303375-1GEA 473 EUR
APOSPC303375-1G
3-Chloro-2-fluoro-4-(trifluoromethyl)aniline 98%
3-Chloro-2-fluoro-4-(trifluoromethyl)aniline
Formel: C₇H₄ClF₄N
Molecular Weight: 213,56 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28752559
CAS-Nummer: 1807135-05-2
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR