2-Fluoro-3-methyl-5-(trifluoromethyl)benzonitrile 97%

Lieferant: Apollo Scientific

PC303382-1G PC303382-250MG
APOSPC303382-1GEA 592 EUR
APOSPC303382-1G APOSPC303382-250MG
2-Fluoro-3-methyl-5-(trifluoromethyl)benzonitrile 97%
2-Fluoro-3-methyl-5-(trifluoromethyl)benzonitrile
Formel: C₉H₅F₄N
Molecular Weight: 203.136 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31916395
CAS-Nummer: 2383627-27-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR