2-Fluoro-3-methyl-5-(trifluoromethyl)benzonitrile 97%
Lieferant: Apollo Scientific
PC303382-1G
PC303382-250MG
APOSPC303382-1GEA
592
EUR
APOSPC303382-1G
APOSPC303382-250MG
2-Fluoro-3-methyl-5-(trifluoromethyl)benzonitrile 97%
2-Fluoro-3-methyl-5-(trifluoromethyl)benzonitrile
Formel:
C₉H₅F₄N Molecular Weight: 203.136 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31916395 CAS-Nummer: 2383627-27-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...