2-Fluoro-3-methyl-5-(trifluoromethyl)benzylamine

Lieferant: Apollo Scientific

PC303386-1G
APOSPC303386-1GEA 1280 EUR
APOSPC303386-1G
2-Fluoro-3-methyl-5-(trifluoromethyl)benzylamine
2-Fluoro-3-methyl-5-(trifluoromethyl)benzylamine
Formel: C₉H₉F₄N
Lagertemperatur: Raumtemperatur
CAS-Nummer: 2383993-46-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR